Assay Design



Gene Transcripts

Gene Symbol
Ube2s
Entrez Gene ID
100774405
Gene Name
ubiquitin-conjugating enzyme E2S
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614505: 188026 - 191087 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648852.1 XP_007647042.1 2 - 3 374 94
GenBank mRNA JI880942.1 - 2 - 3 374 94

View Full Product Details