Assay Design



Gene Transcripts

Gene Symbol
Slc25a29
Entrez Gene ID
100767571
Gene Name
solute carrier family 25 (mitochondrial carnitine/acylcarnitine carrier), member 29
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613993: 1090909 - 1098884 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644296.1 XP_007642486.1 3 - 4 196 62

View Full Product Details