Assay Design



Gene Transcripts

Gene Symbol
Myog
Entrez Gene ID
100751638
Gene Name
myogenin (myogenic factor 4)
Gene Aliases
Myogenin
Chromosome Location
Chr.Un|NW_003613706: 161358 - 163123 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498844.1 XP_003498892.1 2 - 3 550 51

View Full Product Details