Assay Design



Gene Transcripts

Gene Symbol
Cdk2ap2
Entrez Gene ID
100758502
Gene Name
cyclin-dependent kinase 2 associated protein 2
Gene Aliases
I79_018160
Chromosome Location
Chr.Un|NW_003614797: 188440 - 190074 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509929.2 XP_003509977.1 1 - 2 174 103

View Full Product Details