Assay Design



Gene Transcripts

Gene Symbol
Cxcl13
Entrez Gene ID
100773550
Gene Name
chemokine (C-X-C motif) ligand 13
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615544: 54549 - 59215 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512697.2 XP_003512745.1 2 - 3 209 74

View Full Product Details