Assay Design



Gene Transcripts

Gene Symbol
Cited1
Entrez Gene ID
100759164
Gene Name
Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614593: 472448 - 474744 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007649430.1 XP_007647620.1 1 - 2 1140 92

View Full Product Details