Assay Design



Gene Transcripts

Gene Symbol
Mis18a
Entrez Gene ID
100766257
Gene Name
MIS18 kinetochore protein A
Gene Aliases
I79_000808
Chromosome Location
Chr.Un|NW_003613595: 3376195 - 3396389 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642311.1 XP_007640501.1 4 - 5 546 69

View Full Product Details