Assay Design



Gene Transcripts

Gene Symbol
Tenc1
Entrez Gene ID
100752687
Gene Name
tensin like C1 domain containing phosphatase (tensin 2)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617247: 55755 - 72176 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007655053.1 XP_007653243.1 21 - 22 4182 85
GenBank mRNA JI891679.1 - 6 - 7 1219 85
JP037827.1 - 6 - 7 1218 85

View Full Product Details