Assay Design



Gene Transcripts

Gene Symbol
Tenc1
Entrez Gene ID
100752687
Gene Name
tensin like C1 domain containing phosphatase (tensin 2)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617247: 55755 - 72176 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007655053.1 XP_007653243.1 26 - 27 4601 116
GenBank mRNA JP037827.1 - 11 - 12 1637 116

View Full Product Details