Assay Design



Gene Transcripts

Gene Symbol
Tenc1
Entrez Gene ID
100752687
Gene Name
tensin like C1 domain containing phosphatase (tensin 2)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617247: 55755 - 72176 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007655053.1 XP_007653243.1 2 - 3 878 71

View Full Product Details