Assay Design



Gene Transcripts

Gene Symbol
Clec4e
Entrez Gene ID
100773355
Gene Name
C-type lectin domain family 4, member E
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614841: 388046 - 393549 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510181.2 XP_003510229.1 3 - 4 221 66

View Full Product Details