Assay Design



Gene Transcripts

Gene Symbol
Lrwd1
Entrez Gene ID
100766394
Gene Name
leucine-rich repeats and WD repeat domain containing 1
Gene Aliases
I79_007273
Chromosome Location
Chr.Un|NW_003613822: 1001172 - 1006844 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007641688.1 XP_007639878.1 3 - 4 387 87

View Full Product Details