Assay Design



Gene Transcripts

Gene Symbol
Rpl22l1
Entrez Gene ID
100750612
Gene Name
ribosomal protein L22-like 1
Gene Aliases
I79_020083
Chromosome Location
Chr.Un|NW_003615148: 81233 - 83463 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652094.1 XP_007650284.1 3 - 4 260 114
XM_007652095.1 XP_007650285.1 3 - 4 235 114
GenBank mRNA JI869855.1 - 1 - 2 114 114

View Full Product Details