Assay Design



Gene Transcripts

Gene Symbol
Tmcc1
Entrez Gene ID
100771412
Gene Name
transmembrane and coiled-coil domain family 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614360: 481278 - 612411 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507351.2 XP_003507399.1 4 - 5 1816 90
XM_007647939.1 XP_007646129.1 6 - 7 1970 90
XM_007647941.1 XP_007646131.1 5 - 6 1520 90
XM_007647942.1 XP_007646132.1 3 - 4 1348 90
XM_007647943.1 XP_007646133.1 4 - 5 1244 90

View Full Product Details