Assay Design



Gene Transcripts

Gene Symbol
Parp10
Entrez Gene ID
100757141
Gene Name
poly (ADP-ribose) polymerase family, member 10
Gene Aliases
I79_015374
Chromosome Location
Chr.Un|NW_003614411: 516112 - 529479 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648251.1 XP_007646441.1 11 - 12 2811 71
GenBank mRNA JI876917.1 - 2 - 3 373 71

View Full Product Details