Assay Design



Gene Transcripts

Gene Symbol
Cd83
Entrez Gene ID
100769362
Gene Name
CD83 molecule
Gene Aliases
I79_003609
Chromosome Location
Chr.Un|NW_003613670: 978016 - 1015600 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497943.2 XP_003497991.1 2 - 3 305 103

View Full Product Details