Assay Design



Gene Transcripts

Gene Symbol
Nmbr
Entrez Gene ID
100773994
Gene Name
neuromedin B receptor
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613836: 407202 - 415697 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501188.1 XP_003501236.1 1 - 2 419 89

View Full Product Details