Assay Design



Gene Transcripts

Gene Symbol
Rexo4
Entrez Gene ID
100770295
Gene Name
REX4, RNA exonuclease 4 homolog (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615220: 84207 - 95537 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511725.2 XP_003511773.1 5 - 6 1302 68

View Full Product Details