Assay Design



Gene Transcripts

Gene Symbol
Six2
Entrez Gene ID
100767057
Gene Name
SIX homeobox 2
Gene Aliases
I79_001588
Chromosome Location
Chr.Un|NW_003613615: 3265847 - 3268649 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496238.2 XP_003496286.2 1 - 2 444 94

View Full Product Details