Assay Design



Gene Transcripts

Gene Symbol
Ankrd46
Entrez Gene ID
100756375
Gene Name
ankyrin repeat domain 46
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615110: 239773 - 249487 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511432.2 XP_003511480.1 2 - 3 491 80
GenBank mRNA JI893661.1 - 1 - 2 133 80
JP055407.1 - 1 - 2 133 80

View Full Product Details