Assay Design



Gene Transcripts

Gene Symbol
Fam78b
Entrez Gene ID
100760702
Gene Name
family with sequence similarity 78, member B
Gene Aliases
I79_012751
Chromosome Location
Chr.Un|NW_003614161: 282786 - 312303 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007646036.1 XP_007644226.1 2 - 3 195 85

View Full Product Details