Assay Design



Gene Transcripts

Gene Symbol
Scgb1c1
Entrez Gene ID
100770790
Gene Name
secretoglobin, family 1C, member 1
Gene Aliases
I79_025080
Chromosome Location
Chr.Un|NW_003618267: 16785 - 18415 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515510.2 XP_003515558.1 2 - 3 729 74

View Full Product Details