Assay Design



Gene Transcripts

Gene Symbol
Il24
Entrez Gene ID
100773869
Gene Name
interleukin 24
Gene Aliases
I79_003374
Chromosome Location
Chr.Un|NW_003613661: 2946395 - 2949869 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007616626.1 XP_007614816.1 4 - 5 471 64

View Full Product Details