Assay Design



Gene Transcripts

Gene Symbol
Polr2d
Entrez Gene ID
100755604
Gene Name
polymerase (RNA) II (DNA directed) polypeptide D
Gene Aliases
I79_019253
Chromosome Location
Chr.Un|NW_003614974: 155990 - 159373 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007651422.1 XP_007649612.1 2 - 3 395 64
GenBank mRNA JI888655.1 - 2 - 3 395 64

View Full Product Details