Assay Design



Gene Transcripts

Gene Symbol
Hyal3
Entrez Gene ID
100753318
Gene Name
hyaluronoglucosaminidase 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613788: 1246936 - 1253023 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500403.2 XP_003500451.1 3 - 4 1239 117
XM_007641100.1 XP_007639290.1 2 - 3 1007 117
XM_007641101.1 XP_007639291.1 3 - 4 1072 117
GenBank mRNA JI868775.1 - 2 - 3 190 117

View Full Product Details