Assay Design



Gene Transcripts

Gene Symbol
Nr0b2
Entrez Gene ID
100769023
Gene Name
nuclear receptor subfamily 0, group B, member 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614517: 296369 - 299084 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508295.2 XP_003508343.1 1 - 2 544 83

View Full Product Details