Assay Design



Gene Transcripts

Gene Symbol
Tnfrsf13c
Entrez Gene ID
100762567
Gene Name
tumor necrosis factor receptor superfamily, member 13C
Gene Aliases
I79_021096
Chromosome Location
Chr.Un|NW_003615427: 178337 - 179537 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512340.1 XP_003512388.1 2 - 3 364 89

View Full Product Details