Assay Design



Gene Transcripts

Gene Symbol
Lhfpl1
Entrez Gene ID
100753727
Gene Name
lipoma HMGIC fusion partner-like 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614148: 31214 - 84340 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505399.1 XP_003505447.1 2 - 3 478 74

View Full Product Details