Assay Design



Gene Transcripts

Gene Symbol
Tm2d1
Entrez Gene ID
100755825
Gene Name
TM2 domain containing 1
Gene Aliases
I79_003206
Chromosome Location
Chr.Un|NW_003613655: 2490795 - 2519644 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497604.2 XP_003497652.1 6 - 7 674 117

View Full Product Details