Assay Design



Gene Transcripts

Gene Symbol
Fbln7
Entrez Gene ID
100755635
Gene Name
fibulin 7
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613665: 417315 - 438647 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497823.1 XP_003497871.1 4 - 5 570 61

View Full Product Details