Assay Design



Gene Transcripts

Gene Symbol
Htr7
Entrez Gene ID
100764536
Gene Name
5-hydroxytryptamine (serotonin) receptor 7, adenylate cyclase-coupled
Gene Aliases
I79_004939
Chromosome Location
Chr.Un|NW_003613711: 1242684 - 1331686 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498959.2 XP_003499007.1 2 - 3 1186 82

View Full Product Details