Assay Design



Gene Transcripts

Gene Symbol
Vstm2l
Entrez Gene ID
100764618
Gene Name
V-set and transmembrane domain containing 2 like
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613641: 1725086 - 1734081 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007607963.1 XP_007606153.1 3 - 4 243 62

View Full Product Details