Assay Design



Gene Transcripts

Gene Symbol
Cldn11
Entrez Gene ID
100765050
Gene Name
claudin 11
Gene Aliases
Claudin-11
Chromosome Location
Chr.Un|NW_003613976: 143680 - 154892 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503492.2 XP_003503540.1 2 - 3 394 86

View Full Product Details