Assay Design



Gene Transcripts

Gene Symbol
Rasa4b
Entrez Gene ID
100765808
Gene Name
RAS p21 protein activator 4B
Gene Aliases
Rasa4
Chromosome Location
Chr.Un|NW_003613822: 976530 - 993005 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007641687.1 XP_007639877.1 4 - 5 451 63

View Full Product Details