Assay Design



Gene Transcripts

Gene Symbol
Pts
Entrez Gene ID
100750756
Gene Name
6-pyruvoyltetrahydropterin synthase
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613694: 1371272 - 1378752 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498538.2 XP_003498586.1 1 - 2 150 86
GenBank mRNA JP049899.1 - 1 - 2 149 86

View Full Product Details