Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_003500455.2 | XP_003500503.1 | 6 - 7 | 1554 | 72 |
GenBank mRNA | JI886453.1 | - | 2 - 3 | 999 | 72 |
JP044888.1 | - | 1 - 2 | 213 | 72 | |
JP052645.1 | - | 3 - 4 | 1042 | 72 |