Assay Design



Gene Transcripts

Gene Symbol
Tff1
Entrez Gene ID
100764859
Gene Name
trefoil factor 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614068: 773318 - 776585 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007645006.1 XP_007643196.1 3 - 4 248 59

View Full Product Details