Assay Design



Gene Transcripts

Gene Symbol
Exog
Entrez Gene ID
100750872
Gene Name
endo/exonuclease (5'-3'), endonuclease G-like
Gene Aliases
I79_013213
Chromosome Location
Chr.Un|NW_003614199: 334797 - 360553 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505922.2 XP_003505970.1 5 - 6 653 96
GenBank mRNA JI896513.1 - 4 - 5 490 96
JP037634.1 - 4 - 5 490 96

View Full Product Details