Assay Design



Gene Transcripts

Gene Symbol
Znf18
Entrez Gene ID
100768111
Gene Name
zinc finger protein 18
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613651: 2016724 - 2050946 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497497.2 XP_003497545.1 6 - 7 984 103
XM_007613105.1 XP_007611295.1 6 - 7 1137 103
XM_007613121.1 XP_007611311.1 6 - 7 1136 103

View Full Product Details