Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_007652284.1 | XP_007650474.1 | 2 - 3 | 816 | 108 |
XM_007652285.1 | XP_007650475.1 | 3 - 4 | 270 | 108 | |
XM_007652286.1 | XP_007650476.1 | 2 - 3 | 179 | 108 | |
XM_007652287.1 | XP_007650477.1 | 2 - 3 | 208 | 108 | |
XM_007652288.1 | XP_007650478.1 | 2 - 3 | 728 | 108 |