Assay Design



Gene Transcripts

Gene Symbol
Treml4
Entrez Gene ID
100757253
Gene Name
triggering receptor expressed on myeloid cells-like 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615227: 20537 - 36809 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652284.1 XP_007650474.1 3 - 4 1166 77
XM_007652285.1 XP_007650475.1 4 - 5 620 77
XM_007652286.1 XP_007650476.1 3 - 4 529 77
XM_007652287.1 XP_007650477.1 3 - 4 558 77
XM_007652288.1 XP_007650478.1 3 - 4 1078 77
GenBank mRNA JI878795.1 - 1 - 2 170 77

View Full Product Details