Assay Design



Gene Transcripts

Gene Symbol
Treml4
Entrez Gene ID
100757253
Gene Name
triggering receptor expressed on myeloid cells-like 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615227: 20537 - 36809 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652284.1 XP_007650474.1 5 - 6 1264 81
XM_007652285.1 XP_007650475.1 6 - 7 718 81
XM_007652286.1 XP_007650476.1 5 - 6 627 81
XM_007652287.1 XP_007650477.1 5 - 6 656 81
XM_007652288.1 XP_007650478.1 5 - 6 1176 81
GenBank mRNA JI878795.1 - 3 - 4 268 81

View Full Product Details