Assay Design



Gene Transcripts

Gene Symbol
Usp35
Entrez Gene ID
100765174
Gene Name
ubiquitin specific peptidase 35
Gene Aliases
I79_018590
Chromosome Location
Chr.Un|NW_003614860: 432389 - 448319 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007650953.1 XP_007649143.1 5 - 6 592 81

View Full Product Details