Assay Design



Gene Transcripts

Gene Symbol
Kcnj13
Entrez Gene ID
100774709
Gene Name
potassium inwardly-rectifying channel, subfamily J, member 13
Gene Aliases
I79_020263
Chromosome Location
Chr.Un|NW_003615196: 277061 - 280305 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511678.2 XP_003511726.1 1 - 2 701 95

View Full Product Details