Assay Design



Gene Transcripts

Gene Symbol
Dnajc22
Entrez Gene ID
100769451
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 22
Gene Aliases
I79_024102
Chromosome Location
Chr.Un|NW_003616964: 44859 - 48742 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514784.2 XP_003514832.1 2 - 3 1153 99

View Full Product Details