Assay Design



Gene Transcripts

Gene Symbol
Prrg3
Entrez Gene ID
100772670
Gene Name
proline rich Gla (G-carboxyglutamic acid) 3 (transmembrane)
Gene Aliases
I79_018287
Chromosome Location
Chr.Un|NW_003614816: 66517 - 150135 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510039.1 XP_003510087.1 12 - 13 1743 85

View Full Product Details