Assay Design



Gene Transcripts

Gene Symbol
Elk4
Entrez Gene ID
100755459
Gene Name
ELK4, ETS-domain protein (SRF accessory protein 1)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613838: 986211 - 994959 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501205.2 XP_003501253.1 1 - 2 216 132

View Full Product Details