Assay Design



Gene Transcripts

Gene Symbol
Kbtbd8
Entrez Gene ID
100751978
Gene Name
kelch repeat and BTB (POZ) domain containing 8
Gene Aliases
I79_000347
Chromosome Location
Chr.Un|NW_003613584: 5404189 - 5415621 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007615617.1 XP_007613807.1 2 - 3 1374 107

View Full Product Details