Assay Design



Gene Transcripts

Gene Symbol
Plekhm3
Entrez Gene ID
100760707
Gene Name
pleckstrin homology domain containing, family M, member 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614305: 318682 - 454115 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506917.2 XP_003506965.1 4 - 5 2081 60

View Full Product Details