Assay Design



Gene Transcripts

Gene Symbol
Socs6
Entrez Gene ID
100751669
Gene Name
suppressor of cytokine signaling 6
Gene Aliases
I79_016130
Chromosome Location
Chr.Un|NW_003614507: 157000 - 178746 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508249.2 XP_003508297.1 2 - 3 506 101

View Full Product Details